LiquiDispo Disposable Sampler, Bürkle
Proveedor: Bürkle
5393-1120 5393-1131 5393-1130 5393-1121
10821-341PK 6856.08 MXN
10821-341 10821-343 10821-345 10821-309
LiquiDispo Disposable Sampler, Bürkle
Muestreadores Muestreadores de líquidos
LiquiDispo Disposable Samplers provide the highest standards in sampling
  • Ergonomic handle
  • No seals
  • HDPE construction
  • Cleanroom manufactured
  • Gamma ray sterile and nonsterile formats available
The LiquiDispo disposable sampler can be used to take samples of liquids from highly liquid to slightly viscous. An ergonomic handle makes it easier to draw the sample, as with a syringe. Disposable design virtually eliminates the threat of cross contamination.
Unlike conventional reusable samplers, disposable samplers do not require cleaning after use. This eliminates a demanding and often complicated task, with greasy, strongly adhesive, or toxic substances now avoided.
Información para pedidos: LiquiDispo Disposable Samplers are packaged 20 per pack.