STERIL-PEEL™ Self-Seal Sterilization Rolls, GS Medical Packaging
Proveedor: GS Medical Packaging
221436
89201-542CS 8083.94 MXN
89201-542
STERIL-PEEL™ Self-Seal Sterilization Rolls, GS Medical Packaging
Indicadores de esterilización
All STERIL-PEEL™ products are manufactured from medical grade bleached kraft paper
STERIL-PEEL™ products are printed with dual indicators which change color in either steam or ethylene oxide. Both indicators conform with the latest ISO 11140 and EN868 regulations. The steam indicator ink will only change in the presence of all necessary parameters (saturated steam, temperature, and time).
Hydrogen peroxide, rather than chlorine, is used as the bleaching agent. They are made from virgin pulp, ensuring optimum consistency of the paper sheet, with superior barrier properties. All rolls are lead-free and feature medically safe inks.
Certificaciones: 510K and CE compliant.