API® Microbial Identification Kits, Biomerieux
Proveedor: bioMérieux
50300 10400 20800 20210 20500 25200 20900 20900 20210 20500 50300 20800
95060-852PK 9891.91 MXN
CA-95060-690 95060-852 CA-89126-944 95060-692 95060-684 CA-95060-852 CA-95060-684 95060-690 95060-706 95060-668 89126-944 CA-95060-692
API® Microbial Identification Kits, Biomerieux
Tests para microbiología Tests de microorganismos
API® range is the most extensive range available for manual identification of microorganisms
  • Economical
  • Easy to use
  • Rapid identification
It includes a comprehensive panel of identification system covering all groups of bacteria encountered in industrial microbiology .
Additional reagents required see package insert for details
https://us.vwr.com/store/product/18630970/microbial-identification-reagents-biomerieux
API Web software required for Identification -purchase directly from bioMerieux.