Compact Titrators, METTLER TOLEDO
Proveedor: Mettler-Toledo
B39950001M 30267116 30267114 30252666 30252669 30252671 30267117 30252661 30267115 30252660
10861-688EA 585810 MXN
10861-676 10861-688 10861-678 10861-694 10861-684 10861-674 10861-686 10861-690 10861-692 75853-862
Compact Titrators, METTLER TOLEDO
Valoradores
METTLER TOLEDO Compact Titrators are designed for routine applications and simple operation.
The large color touchscreen with the One Click user interface is easy to understand and operate. The touchscreen is optimized for direct access to routine tasks and provides clear information to the user.
Security is an important concept in all METTLER TOLEDO Titrators. Many parameters are automatically checked by the instrument, allowing you to keep full control of the titration analysis. Connectivity to LabX Titration Software allows for security and efficient management of results.
The Karl Fischer Titrators combine fast and precise water content determination with very simple operation. The G10S and G20S are ideal for potentiometric titration with their easy-to-use interface and space saving design; they are the optimal choice for your daily routine analysis.