PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Proveedor: DWK Life Sciences
73818-24 73818-20 73818A-24 73818-15 774161-0020 73818X-24 774161-0015 73818X-18 774161-0924 73818-18 774161-0008 774161-0024 774161-0018 774161-0013 N73818T-13
66010-751CS 3783.09 MXN
KT774161-0008 KT774161-0924 93001-096 66010-751 KT774161-0015 KT774161-0018 66010-796 66010-774 66010-785 66010-946 66010-945 66010-947 KT774161-0024 KT774161-0013 KT774161-0020
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Septos
Ideal for use with open-top caps to access container contents with a syringe.
  • Resists coring
  • Highly chemically-resistant PTFE facing maintains integrity of content
  • Silicone rubber backing allows repeated puncturing through the seal
  • Recommended for use in all autosamplers using screw thread vials