Order Entry
VWR® Animal Ear Punches
VWR® Animal Ear Punches
  10806-290
undefined
VWR® Animal Ear Punches
  10806-290
$994.30
Select uom
Unit of Measure
Each (1UN)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Ø:
    2 mm Hole Size
  • Description:
    Animal Ear Punch, Plier-Style
  • Length:
    10.8 cm (4.25")
  • Material:
    Stainless steel, surgical grade
  • Cat. no.:
    10806-290
VWR® Animal Ear Punches are made from corrosion-resistant ASTM (899-94) Series 400 stainless steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These ear punches are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.