Support Rings with Clamp, Chemglass
Proveedor: Chemglass
CG-3057-12 CG-3057-11
10034-450EA 1445.48 MXN
10034-450 10034-448
Support Rings with Clamp, Chemglass
Anillos de retorta
Solid ring with built in clamp for attachment directly to support rod
Made of Nickel plated zinc alloy.