HazMat Mat Pad, PIG®
Proveedor: New Pig
MAT3510 MAT310
56620-690PK 3600 MXN
56620-690 10860-084
HazMat Mat Pad, PIG®
Materiales para contención de derrames
The HazMat Mat Pad absorbs the widest range of acids, bases, corrosives, chemicals and unknown liquids.
  • Chemical-resistant mat won't degrade
  • Won't rip, tear or fray even when saturated
  • Absorbs high concentrations like 98% sulfuric acid and 30% sodium hydroxide
Absorbs the widest range of acids, bases, corrosives, chemicals and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated.