Disposable Samplers, PowderDispo, Burkle
5393-5522 5393-5510 5393-5511
10821-288 10821-322 10821-290
Disposable Samplers, PowderDispo, Burkle
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances
  • HDPE
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Disposable sampler for the highest standards
A complete all-layer sample can be taken from bags or smaller containers with the lance.