SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Proveedor: SP Industries
LG-4241T-100 LG-4241T-104 LG-4241T-102
14208-260EA 10248.94 MXN
14203-812 14208-260 14205-414
SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Buretas
This inexpensive portable titration set features a 2mm straight stopcock with PTFE plug
The borosilicate glass burette is filled by pressure supplied by the rubber bulb. A small hole in the filling tube at the "O" line permits setting of the liquid level and excess titrant is returned to the bottle. Complete Set furnished with burette, bottle, rubber hose, rubber stopper, and rubber bulb.