Reagent Reservoirs, VistaLab
Proveedor: VistaLab
3054-2001 3054-1010 3054-5000 3054-2002 4054-1010 3054-1006 3054-1007 3054-1003 3054-2006 3054-2007 3054-2000 3054-2012 3054-3000 4054-1006 3054-1004 3054-1005 3054-2003 3054-1012 3054-3001 4054-1012 3054-1011 3054-1002 3054-1000 3054-1001 4054-1004 4054-1000 4054-1002 3054-2013 4054-1011 3054-1013
89003-382CS 3225.05 MXN
41428-960 89003-694 10770-298 10770-294 10770-296 14007-065 14007-066 10770-292 75813-820 14007-069 75813-806 89003-382 75814-382 41428-950 75814-380 89003-384 41428-952 41428-954 41428-956 10770-300 41428-958 14007-076 14007-074 14007-072 14007-070 93000-412 93000-410 75813-814 75813-816 75813-818
Reagent Reservoirs, VistaLab
Depósitos de reactivos
VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
  • Trough-within-a-trough design for greatest sample recovery
  • Pour-off spouts at every corner
  • Wider base for stability
  • Free of animal products
  • Sterile reservoirs are RNase/DNase-free and non-pyrogenic
  • Bulk or individually wrapped
  • 12-channel polypropylene reservoir is autoclavable
Sterile and non-sterile reservoirs and reservoir coolers are available for purchase, with bulk or individually wrapped formats for compatibility with any distribution method.