Drying/Heating Ovens with Forced Convection, Avantgarde.line FD and FED Series, BINDER
Proveedor: Binder
9010-0304 9010-0300 9010-0306 9010-0310 9010-0296 9010-0294 9010-0302
75783-610EA 7738.56 MXN
75783-610 75783-602 76413-498 75783-598 75783-594 75783-606 75783-590
Drying/Heating Ovens with Forced Convection, Avantgarde.line FD and FED Series, BINDER
Hornos Hornos de convección natural y con ventilación
Forced convection ovens are ideal for sterilization, drying processes, heat treatments, thermal storage, and testing.
  • Precise heating results
  • Easy to clean
These ovens maintain uniform temperature distribution and symmetrical airflow throughout the entire inner chamber for precise heating results
The short recovery time after door opening decreases temperature fluctuation. The minimal heat-up time and high air transfer rate ensure fast, intensive drying performance; especially on materials with high moisture content. The Advanced Preheating Technology (APT.line™) provides temperature accuracy of 1°C with a wide temperature range from 10°C above ambient to 300 °C. With inner chamber constructed of stainless steel with rounded edges and internally welded seams they are easy to clean. The completely tempered interior prevents condensation and ensures moisture deposits will not impair drying results.
The 50mm (2") diameter motorized exhaust duct with 5 available positions programmable from the controller allows for quick vapor elimination preventing condensation in the inner chamber. A powerful fan is provided for air circulation. In FED models a powerful yet quiet, digitally adjustable air turbine provides increased air circulation.
High-quality insulation reduces heat dissipation, contributing to an active reduction in ambient temperature; removing the need for additional air conditioning in the lab. The low external housing temperature (50°C maximum) provides protection from burns. The integral thermostat is rated Class 2, according to DIN 12880, Part 1.
All models include a USB port for data recording and an optional Ethernet interface.
Certificaciones: UL.
Información para pedidos: All ovens include two chrome-plated shelves. FED series include an RS-422 interface port. Connecting the oven to a printer or PC through this port requires an interface converter; contact your Avantor representative for more information. For additional sizes not listed below, please refer to the BINDER Classic FD/FED Drying Ovens.