Microbiological Incubators, Avantgarde.line BF Series, BINDER
Proveedor: Binder
9010-0320 9010-0313 9010-0316 9010-0314 9010-0319 9010-0315
75783-546EA 5313.6 MXN
75783-612 75783-546 75783-550 75783-614 75783-548 75783-616
Microbiological Incubators, Avantgarde.line BF Series, BINDER
Incubadoras Incubadores estándar
BINDER microbiological incubators are ideal for research, production and quality assurance.
  • APT.Line™ preheating chamber
  • Microprocessor PID controller
These incubators feature the APT.Line™ preheating chamber for reliable temperature accuracy and uniformity in applications requiring constant, reproducible incubation conditions
Units have an inner tempered safety glass door, corrosion resistant stainless steel chamber and two chrome plated racks. They also carry a Class 3.1 independent temperature safety device with visual and audible temperature alarm.
Incubators are equipped with a microprocessor PID controller with a 2 line LCD display. The timer can be set for up to 99 days, 99 hours, 59 minutes for continuous operation. The unit also contains an integrated timer, and one ramp function. They also feature a USB port for data recording and an optional Ethernet interface.
The BF 56, and BF 115 models are stackable.
Certificaciones: UL.