VWR® Beakers with Handle, PP
Proveedor: VWR
VWR®
431090 431060 431040 431070 421071 431080 431050
75800-256CS 554.95 MXN
75800-256 75800-258 75800-260 75800-262 75800-250 75800-252 75800-254
VWR® Beakers with Handle, PP
Vasos de precipitados
VWR® polypropylene beakers, which come in a natural color, have a handle, pouring spout and graduations printed directly on the beakers.
  • Autoclavable
  • Integrated pouring lip
  • Natural color
  • Printed graduation
The beakers are autoclavable. Working temperatures of 0 to 121 °C.
Certificaciones: Resin meets US FDA 21 CFR requirements.
Precaución: Do not stack products while autoclaving.