Infrared Lamp and Reflector
470005-592
: Avantor
- Description:Infrared lamp and reflector, complete set
- Cat. no.:470005-592
- Supplier No.:WARD470005-592
About this item
Comes with 125 W clear, heat bulb.
- Color temperature: 2500 K
Comes with 125 W clear, heat bulb.