RO Series Multi-Position Stirrer Plates, IKA Works
Proveedor: IKA
0003691000 0003690500 0003692500
89426-972EA 2602.8 MXN
89426-972 89426-970 89426-968
RO Series Multi-Position Stirrer Plates, IKA Works
Imánes de agitación Agitadores magnéticos multiposición con placas calefactoras
5, 10, or 15-position digital magnetic stirrer without heating, designed for synchronous stirring
The magnetic coil technology provides noiseless and consistent stirring on all positions. Speed remains constant, even when load changes. The closed and compact design allows easy cleaning and protects the equipment against the penetration of liquids. Additionally, a removable transparent anti-slip mat is included.
Certificaciones: UL, cUL, CSA, CE certified, RoHS compliant.
Información para pedidos: Unit comes with detachable power cord. Three year mnaufacturer's warranty with product registration.