Wool Fire Blanket with Cylindrical Wall Case
Proveedor: COTTON GOODS MFG CO INC SE
FR25802C
470122-236EA 3558.92 MXN
470122-236
Wool Fire Blanket with Cylindrical Wall Case
Suministros de primeros auxilios Mantas contra incendios
Perfect Fire Aid Blanket for Any Laboratory
  • Gray fire blanket
  • 80% wool, 20% man-made fiber
  • Wall Case: 9" Ø×19.5" L
Flame resistant wool blend blanket is designed for smothering fires. It can also be used as a First Aid blanket. The convenient wall case design includes brackets for wall mounting and provides easy accessibility in the lab.