Orion™ Star™ A111 pH Benchtop Meter, Thermo Scientific
Proveedor: Thermo Scientific
STARA1117 STARA1116 STARA1115 STARA1110
89206-278EA 20082.84 MXN
76076-752 89206-278 89260-358 89206-280
Orion™ Star™ A111 pH Benchtop Meter, Thermo Scientific
Medidores electroquímicos Medidores de pH
Take simple pH, mV and temperature measurements with the Orion™ Star™ A111 pH Benchtop Meter, which offers easy operation for basic laboratory testing.
  • AUTO-READ™ locks in the stable reading on screen and a ready indicator alerts when readings are stable
  • Meter offers up to three point pH calibration and automatic recognition of USA/NIST and DIN buffers
  • Easily recall calibration slope data for procedure checks
  • Non-volatile memory holds up to 50 data points
  • IP 54 rated housing can handle splashes and can be wall-mounted
Take simple, routine pH, mV, and temperature measurements with the Orion™ Star™ A111 pH Benchtop Meter, an easy-to-use and budget-friendly instrument for basic pH analysis in the laboratory.
Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information, and power source. Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples. Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
Perform up to a three point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Certificaciones: CE, TUV 3-1, FCC Class A.
Información para pedidos: For compatible pH electrodes, see Orion™ items 9157BNMD and GD9156BNWP.