Orion™ Versa Star Pro™ pH/Conductivity Multiparameter Benchtop Meter, Thermo Scientific
Proveedor: Thermo Scientific
VSTAR50 VSTAR52
89206-400EA 78448.85 MXN
89206-398 89206-400
Orion™ Versa Star Pro™ pH/Conductivity Multiparameter Benchtop Meter, Thermo Scientific
Medidores electroquímicos Medidores de pH
Get premium performance and flexibility with the Orion™ Versa Star Pro™ Meter with pH and Conductivity Modules for simultaneous pH, conductivity, and temperature analysis.
  • Onscreen measurement stability indicator lets users know exactly when measurements are ready
  • Auto-Read mode holds a stable reading, Continuous mode shows changing readings, Timed mode logs data at specific intervals, and Single-Shot mode holds a reading after a preset time is reached
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
  • Easily access, update and switch preferred measurement settings, using ten password-protected methods per channel
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position
  • Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID, and account ID
  • Three-year meter and module warranty
Get high accuracy, premium performance, and ultimate flexibility with the Orion™ Versa Star Pro™ Benchtop Meter with pH and Conductivity Modules.
Versa Star Pro™ benchtop meters are designed to meet the most challenging applications with ease. Delivering rich functionality and features, users can optimally configure the meter system to meet changing requirements. These meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad. Ensure measurement integrity by viewing the active calibration log while taking readings. Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values. Perform up to a six point conductivity calibration with calibration editing option that allows individual points to be fixed without a full recalibration. Calibration editing option allows individual points to be fixed without a full recalibration. Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate. Use linear, non-linear, EP (USP), or off conductivity temperature compensation with 5, 10, 15, 20 or 25°C reference temperatures. Versa Star Pro™ meters offer a system access feature for administrative and custom user access levels to various meter functions including data/time settings, meter setup, data log, and calibration log. Non-volatile meter memory preserves data and settings, even in the event of power loss. IP54-rated housing resists dust and splashes, and can be wall-mounted.
Meet the most challenging applications for pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity, and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. View pH and conductivity simultaneously or view each channel separately. Log up to 2000 data point sets with time/date stamp and easily transfer logs to a printer or computer.
Certificaciones: CE, TUV 3-1, FCC Class A.
Información para pedidos: For compatible pH electrode, see Orion™ item 8157BNUMD. For compatible conductivity probe, see Orion™ item 013005MD. For compatible buffer solutions, see Orion™ items 810199 and 11007.