Difco™ Mueller Hinton Agar, BD
Proveedor: BD Life Sciences
Difco™
225220 225250 225250 225220
90001-822EA 4995.75 MXN
90001-822 CA90001-822 90001-816 CA90001-816
Difco™ Mueller Hinton Agar, BD
Medios Medios preparados
Mueller Hinton Agar is used for the determination of susceptibility of microorganisms to antimicrobial agents.
  • Meat infusion and casein provide nitrogen and other essential nutrients
  • Starch hydrolysis provides dextrose as an energy source
  • Agar is the solidifying agent
Mueller Hinton Agar is used as a test medium for antimicrobial susceptibility testing. Mueller Hinton Agar is recommended for the diffusion of antimicrobial agents impregnated on paper disc through an agar gel as described in CLSI Approved Standard.