AlphaTec® 37-646 General purpose gloves, Ansell
Proveedor: Ansell
AlphaTec®
184701 184702 184700 184703 113147 184704
75795-298CS 9870.34 MXN
75795-298 75795-300 75795-294 75795-296 75795-290 75795-292
AlphaTec® 37-646 General purpose gloves, Ansell
Guantes
An ideal glove for chemical protection with good grip, fit, and comfort.
  • Good chemical, snag, puncture, and abrasion resistance
  • Excellent grip on wet and dry surfaces
  • Form-fitting fingers and a tapered wrist for a more comfortable, ergonomic fit
  • High performance nitrile compound for enhanced flexibility and dexterity, even in cold environments
  • Latex free formulation: no risk of Type I allergies
  • Unflocked: to prevent the risk of food contamination
  • Offer superior mechanical properties over natural rubber gloves
Ansell UltraGrip™ technology features an innovative raised diamond grip to offer unmatched grip in wet and fatty conditions.
Certificaciones: Complies with FDA food handling requirements (21 CFR 177.2600).
Envase: Gloves are packaged 12 pairs in a bag; 144 in a carton.