F Series Recirculating Chillers, JULABO
Proveedor: Julabo
9620050.02 9620100.02 9620050 9620025 9620025.02
89421-862EA 181561.77 MXN
89421-862 CA89421-862 89428-200 CA89428-200 10790-512
F Series Recirculating Chillers, JULABO
Termostatos con circulación Sistemas de circulación cerrada Circuladores de enfriamiento
JULABO F Series chillers for basic laboratory cooling tasks. Ideal for replacing tap water. Provide recirculating fluid to applications at a consistent temperature and pressure. Compact design with easy to use digital interface including a timer. May be used with water or glycol/water mixtures. High quality construction with stainless steel components in combination with German engineering and quality for a long product lifetime.
  • German engineering and quality
  • Stainless steel construction
  • Integrated timer
JULABO F Series environmentally friendly recirculating coolers save tap water and provide powerful cooling for short cool-down times.
The F model series includes three models with 250 W, 500 W or 1 kW of cooling control power at 20 °C with an operating range of 0 to 40 °C or −10 to 40 °C. Julabo F models are an inexpensive way to replace tap water and ideal for basic cooling tasks. Easy operation and compact design make them perfect for cooling rotary evaporators, condensers, Soxhlet apparatus, Kjeldahl instruments, polarimeters and refractometers.
Models feature centrifugal pump, splash-proof membrane keypad with LED temperature display, straightforward filling and draining, filling level indicator, timer and internal float switch.
F250 supplied with 8 and 10 mm barbed fittings. F500 and F1000 include 8 and 12 mm barbed fittings. May be used with water, water/glycol and JULABO Thermal EG fluid.
Due to the compact design, the coolers fit on or under lab benches.
Certificaciones: CE, F250 NRTL for C/US.
Información para pedidos: Accessory castor platform is available for F250 (sold separately).
Información de suministro: Units ship via air ride.