Visio-tek™ Wireless Digital Microscope, Visio-tek
VISIO-UVWIR-US VISIO-UVW-US VISIO-HM-US VISIO-L-US VISIO-P-US VISIO-S-US
75841-170EA 15479.04 MXN
75841-170 75841-166 75841-174 75841-158 75841-156 75841-168
Visio-tek™ Wireless Digital Microscope, Visio-tek
Visio-tek™ is a line of versatile and portable, digital microscopes that utilize both Wi-Fi and USB transmission.
  • Wi-fi
  • Portability
  • High definition microscopy lens
  • Windows (7-8-10), Mac, Andoroid and iOS compatible
  • Free dedicated basic SW and app
  • Led IR and UV models
Visio-tek can be connected to iOS and Android smartphones or tablets via the Visio-tek App, as well as through a Wi-Fi/USB connection to Windows and Mac computers or laptops
Using Visio-tek dedicated software, you can capture pictures, shoot videos, and take measurements. This is the state-of-the-art way to observe the micro-world! The standard model is suitable for multipurpose applications which allows you to see the natural and visible spectrum of light.
The polarised model incorporates polarizing light to decrease glare on plastic, glass, metals, minerals, and paper. The polarizer gives you a darker, more saturated, sharper image with increased contrast.
Model VISIO-UVW is a microscope suitable for special applications requiring UV light: Mineralogy, Graphology, Forensics, Industry, Beauty & Trichology, Inks, Arts, Color and Printing industries.
Model VISIO-HM for applications requiring the greatest magnification: Biology, Botany, Mineralogy, Hair treatment, Textiles.
Model VISIO-UVWIR with IR light, often used in the field of handwriting/graphology.
Certificaciones: CE Compliant. FCC Compliant.