Whatman™ Solvent IFD In-Line Filter/Degassers
Proveedor: Whatman products (Cytiva)
Whatman™
6725-5002A 6725-5002 6725-5002 6725-5002A
28137-971EA 210.38 MXN
CA11013-008 28137-971 CA28137-971 10035-492
Whatman™ Solvent IFD In-Line Filter/Degassers
Filtros Filtros en línea
Cytiva's Solvent IFD is a high-flow HPLC in-line filter/degasser, designed to simultaneously filter and degas organically based HPLC mobile phases.
  • Faster than traditional mobile phase preparation
  • Chemically resistant polypropylene membrane and housing, built to filter solvent mobile phases without expensive stand-alone degassing equipment
  • Air vent on inlet with luer lock cap
  • Integrity-testable with the bubble point method
  • End fittings included
Whatman IFD options from Cytiva's business are available for a variety of applications.
A sturdy and chemically resistant medium is needed to filter and degas an organic solvent. Polypropylene is largely inert, compatible with many chemicals, and retains stability across a range of temperatures. The 0.2 µm high-flow polypropylene membrane and housing makes Solvent IFD suitable to filter and degas these solvents.
The Solvent IFD’s all-polypropylene housing is sealed by a security ring. The filter includes fittings to accommodate ¹/₁₆ to ¹/₈" tubing and an air vent on the inlet with luer lock cap to enable priming.
The in-line filter works on the principle of 'bubble point' – the point of pressure at which gases will pass through a wet membrane. If pressure is maintained below the bubble point, the gas will not pass through the membrane and is trapped by the particular filter device.