Aneroid Barometer
Proveedor: UNITED SCIENTIFIC SUPPLIES
ANBR01
470221-934EA 1718.08 MXN
470221-934
Aneroid Barometer
Barómetros
Instrument for measuring pressure.
  • Ring for easy hanging
  • Aligned with GLOBE protocols
Mounted on a plastic base. The aneroid barometer includes an instruction sheet.