Histo Plas™ Capsettes Histology Cassettes, Bio Plas, Inc
Proveedor: Bio Plas
6042 6037 6030 6036 6040 6031 6033 6043 6041 6034 6035 6032 6038 6039
21150-014CS 13650.25 MXN
21150-014 21150-036 21150-024 21150-034 21150-022 21150-018 21150-028 21150-038 21150-016 21150-026 21150-032 21150-020 21150-030 21150-040
Histo Plas™ Capsettes Histology Cassettes, Bio Plas, Inc
Casetes para histología Casetes para procesamiento de tejidos
Histo Plas™ Capsettes™ provide a modular, organized means for safe and consistent processing, embedding, sectioning and storage of specimen samples.
  • Engineered to eliminate Chatter during sectioning
  • Premium quality
  • Made in the USA
  • Comes in 16 colors
  • Will not open during processing
Histo Plas™ Capsettes™ provide a modular, organized means for safe and consistent processing, embedding, sectioning and storage of specimen samples
Cost effective, yet quality engineered to remain sealed during processing. Strategically designed slots allow for efficient solvent flow and drainage, Histo Plas Capsettes will not float during processing. Our rigid design eliminates undersized cassettes during sectioning. Proprietary acetyl copolymer plastic is utilized to resist heat, solvents, decalcifying solutions, and problems associated with microwave processing. Histo Plas Capsettes will fit standard stainless steel, and disposable plastic base molds.
Available in 16 colors which provide a convenient means of separating surgical and autopsy cases, institutions, pathologist, etc. Histo Plas Capsettes are accepted by all of the current automatic cassette labelers.