Difco™ Letheen Agar, BD Diagnostics
Proveedor: BD Diagnostics
268010 268010
90003-798EA 6242 MXN
90003-798 CA90003-798
Difco™ Letheen Agar, BD Diagnostics
Medios Medios preparados
For evaluating the bactericidal activity of quaternary ammonium compounds.