BLAUBRAND® Bulb Pipettes, 1 Mark, Class AS, USP, Certified, BrandTech
Proveedor: BrandTech
BLAUBRAND®
30613 30618 30612 30603 30619 30611 30608 30602 30606 30615 30605 30614 30617 30610 30616 30609 30601 30607
10218-662PK 1695 MXN
10218-680 76313-890 10218-682 10218-660 10218-670 10218-662 10218-650 10218-672 10218-664 10218-652 10218-674 10218-666 10218-654 10218-676 10218-668 10218-656 10218-678 10218-658
BLAUBRAND® Bulb Pipettes, 1 Mark, Class AS, USP, Certified, BrandTech
Pipetas Pipetas graduadas
Class AS volumetric single mark pipettes.
  • Certified Class A
  • Calibrated to deliver (TD)
  • Constructed according to DIN EN ISO standards with error limits that meet the specifications of USP/ASTM
  • Each individual piece is marked on a computer controlled, automated production line to ensure maximum precision
  • USP batch certificates are supplied
  • Volumetrically stable even when heated up to 250 °C via special annealing process
  • Durable, high-contrast blue enamel markings
BLAUBRAND® single mark bulb pipettes are individually marked to provide Class A 'to deliver' measurements according to USP. Pipettes are made of AR-GLAS® according to the DIN EN ISO standard with error limits that meet USP/ASTM standards. Glass is annealed for durability, and volumetric stability, even when heated to 250˚ C. BLAUBRAND markings of blue enamel are highly legible for precise meniscus setting with a variety of liquids.
Certificaciones: Class AS per DIN EN ISO 648. Meets standards of USP Section 31. AQL of 0.4 per DIN ISO 3951.