Orion™ pH Electrode Filling Solution, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
900011 900002 610011 810007 910008-WA 900004 900019 900003 510011 900001
34108-008PK 2521.81 MXN
34108-019 34108-008 34111-850 34107-413 14004-258 34108-020 34107-806 34111-830 34107-990 14231-146
Orion™ pH Electrode Filling Solution, Thermo Scientific
Electrolitos Electrolitos de relleno
Use Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.
Only use ROSS filling solution with ROSS™ and ROSS Ultra™ pH electrodes to prevent damage. When the single junction reference electrode or double junction reference electrode are used with Orion™ ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide.