Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Proveedor: Honeywell Research Chemicals
HYDRANAL™
34693-10G 34813-500ML 34813-6X500ML 34696-6X25G 34748-6X10G 34446-40ML 34847-6X40ML 34694-80ML 34849-6X80ML 34828-6X40ML 34813-6X100ML 34748-10G 34813-100ML
BJ34748-10GCS 22024.37 MXN
BJ34748-10G BJ34813-6100ML BJ34693-10G BJ34813-100ML BJ34813-500ML BJ34813-6500ML BJ34446-40ML BJ34696-25G BJ34847-40ML BJ-34694-80ML BJ34828-40ML BJ34748-6X10G BJ34849-80ML
Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Normativas Estándares de determinación de agua (según el método Karl Fischer)
Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application.
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty
  • Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Long shelf life (up to five years)
  • Convenient packaging
Información de suministro: Supplied with detailed instruction for use.
Precaución: The recommended storage temperature for product HYDRANAL™ Water Standard is 2-8°C. The product can be transported at normal ambient temperatures. In the long term, temperatures > 20°C (if possible > 8 °C) should be avoided.