Whatman™ Anotop Syringe Filters, Sterile, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™ Anotop™
6809-2122 6809-2112 6809-1102 6809-2102 6809-2122 6809-2112 6809-1102 6809-1112 6809-1122 6809-1112 6809-2102 6809-1122
28138-021EA 759.07 MXN
28138-021 28138-011 CA28138-023 28138-025 CA28138-021 28138-023 CA28138-011 28138-007 CA28138-025 CA28138-009 28138-009 CA28138-007
Whatman™ Anotop Syringe Filters, Sterile, Whatman products (Cytiva)
Filtros Filtros de jeringas
Whatman Anotop sterile syringe filters from Cytiva contain an inorganic Anopore membrane. The low protein binding exhibited by this sterile membrane filter make it well suited for biological filtration.
  • Sterile syringe filters for critical applications
  • Aluminum-based inorganic Anotop syringe filters in three pore sizes
  • Compatible with most organic solvents and aqueous samples
  • Low hold-up volume (< 20 μl for Anotop 10) for maximum sample recovery
  • Made without wetting agents or adhesives for low risk of sample contamination
  • Capillary pore structure supports highly consistent filtration results
  • Recyclable after use
Whatman Anotop filters offer chemical inertness and functionality in a wide range of solvents and temperatures. Sterile Anotop filters can be used as a phage and virus filtration membrane, or for cold sterilization* of growth media, protein removal, filtration prior to spectrophotometric analysis, or other analytical methods.
Unlike many organic membranes, the inorganic Anopore membranes in Anotop syringe filters have sharply defined capillary pore structures with narrow pore-size distribution.
The syringe filter’s pigment-free polypropylene housing is made without wetting agents or adhesives, designed for low extractable levels, low protein binding, and minimal sample hold-up. These features make Anotop syringe filters a good option for filtration of valuable, proteinaceous samples where minimal loss is required.