Hose Tubing Connectors, Bürkle
Proveedor: Bürkle
8700-1416 8700-0810 8700-0406 8700-1012 8700-0608 8700-1214
75876-094PK 474.15 MXN
75876-094 75876-092 75876-090 75876-100 75876-098 75876-096
Hose Tubing Connectors, Bürkle
Racores y conectores
Conical, straight hose connectors for various hose sizes.
  • Polypropylene construction
  • Sterilizable
Envase: 10 connectors per package.