Glassine Weighing Paper Sheets, Dyn-A-Med
Proveedor: Dyn-A-Med Products
80049 80048 80047
490007-234PK 455.52 MXN
490007-234 490007-236 490007-238
Glassine Weighing Paper Sheets, Dyn-A-Med
Papeles de pesaje
Glassine balance weighing paper for analytical weighing is designed for direct placement on any lab balance pan or for transferring material to another container.
  • Lightweight paper holds most analytical samples
  • Smooth, translucent surface will not trap material
  • Clean, dust-free sheets
  • Will not absorb moisture