Esta buscando: Soportes de pipetas
26  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"26"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Numero del catalogo: (470311-176)
Proveedor: VWR International
Descripción: Customize your storage of pipettes.
Numero del catalogo: (82010-756)
Proveedor: VWR International
Descripción: Acrylic, four-place stand provides efficient, convenient storage for most major brands of pipettors.
Numero del catalogo: (82007-220)
Proveedor: VWR International
Descripción: Durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
Numero del catalogo: (82024-536)
Proveedor: VWR International
Descripción: These four-place acrylic pipette racks keep instruments secure and conveniently accessible in the laboratory environment
Numero del catalogo: (89511-786)
Proveedor: VWR International
Descripción: This unique pipettor holder allows users the option of use on the benchtop, or hanging under a shelf to maximize available working space
Proveedor: VWR International
Descripción: Convenient three-, four-, and six-place stands are designed to hold most major brands of pipettors.
Proveedor: VWR International
Descripción: Protects and organizes glass and plastic pipettes.
Proveedor: VWR International
Descripción: Linear or carousel stand for up to six single channel pipettes.
Numero del catalogo: (89140-276)
Proveedor: VWR International
Descripción: The Cargo Manual pipette Rack clicks together horizontally with side connections, and is then securely locked in place with top connections.
Numero del catalogo: (82007-226)
Proveedor: VWR International
Descripción: Autoclavable polypropylene holders keep pipettors at your fingertips.
Proveedor: Bel-Art Products, a Part of SP
Descripción: This polypropylene system is strong enough to support the weight of even the heaviest pipettor.
Numero del catalogo: (53577-017)
Proveedor: Bel-Art Products, a Part of SP
Descripción: Support rack horizontally holds 12 volumetric or graduated Pipettes 1 mL and larger.
Numero del catalogo: (53577-145)
Proveedor: Bel-Art Products, a Part of SP
Descripción: Store pipettors while protecting pipettor tips in these epoxy-coated aluminum racks that provide quick, easy access for more efficient work.
Numero del catalogo: (53576-207)
Proveedor: Bel-Art Products, a Part of SP
Descripción: Holds 50 pipettes in vertical position in 5x10 arrangement.
Proveedor: Bel-Art Products, a Part of SP
Descripción: Diseñadas para el almacenamiento seguro y ordenado de hasta 28 pipetas.
Numero del catalogo: (53577-137)
Proveedor: Bel-Art Products, a Part of SP
Descripción: Unique polypropylene and PVC stand holds up to 94 pipettes and rotates on a central vertical axis for convenient pipette selection.
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 16 of 26
no targeter for Bottom