VWR® Disposable Plastic Transfer Pipette, Graduated
Proveedor: VWR
VWR®
201C.US 200CS20 200CS01 200C.US 218C.US 212C.US
10754-264BX 2237.6 MXN
10754-264 10755-252 10754-268 10754-266 10754-272 10754-270
VWR® Disposable Plastic Transfer Pipette, Graduated
Pipetas Pipetas de transferencia (Pasteur)
VWR® Disposable Transfer Pipettes are designed to safely transfer and dispense liquids in all types of laboratories.
  • Pipettes are packaged using medical grade paper and plastic film
  • Non-toxic pipettes can be safely incinerated or heat-sealed and frozen at temperatures as low as –70 °C (–94 °F)
VWR® pipettes are produced using an optimal blend of low density polyethylene, allowing for highly controlled liquid dispensing.