VWR® Vacuum Desiccator, Bottom and Lid with Socket Valve
Proveedor: VWR
VWR®
1632415287254 1632415287304 1632415287154 1632415287104 1632415287204
75871-430CS 3877.06 MXN
75871-428 75871-430 75871-432 75871-434 75871-436
VWR® Vacuum Desiccator, Bottom and Lid with Socket Valve
Desecadores Desecadores de vidrio
The VWR® Vacuum Desiccator is made from high quality Borosilicate 33 expansion glass
  • Very good chemical resistance
  • High thermal shock and pressure resistance, working with vacuum
  • Autoclavable
It is designed for a variety of vacuum applications, including desiccation of samples and storage of materials sensitive to humidity.