VWR® Desiccator, Bottom and Lid with Glass Knob
Proveedor: VWR
VWR®
1632415287253 1632415287103 1632415287153 1632415287303 1632415287203
75871-658CS 1256.27 MXN
75871-658 75871-660 75871-662 75871-664 75871-666
VWR® Desiccator, Bottom and Lid with Glass Knob
Desecadores Desecadores de vidrio
This VWR® Desiccator is made from high quality Borosilicate 33 expansion glass
  • Very good chemical resistance
  • High thermal shock and pressure resistance, working with vacuum
  • 2 mm tolerance
  • Autoclavable
It is designed for a variety of vacuum applications, including desiccation of samples and storage of materials sensitive to humidity.