SP Wilmad-LabGlass Allihn Condensers, SP Industries
Proveedor: SP Industries
LG-4680-130 LG-4680-102 LG-4680-106 LG-4680-104 LG-4680-100
14204-218EA 4269.54 MXN
14204-860 14204-346 14204-218 14206-490 14204-648
SP Wilmad-LabGlass Allihn Condensers, SP Industries
Condensadores
These borosilicate glass Allihn reflux condensers have outer Standard Taper joint at top and an inner drip Standard Taper joint of the same size at bottom.