Dissolution Vessel Covers, Quality Lab Accessories
11010-174
- Color:Clear
- Description:Hinged Cover for Distek Baths
- Cat. no.:11010-174
- Supplier No.:COVERD-01
QLA manufactures a wide selection of vessel covers for most dissolution testers.
- All covers are made in the USA
- Custom covers are available
The loss of media due to evaporation can be considerable. Vesssel covers reduce these issues and provide openings for cannula, cannula stoppers, temperature verification, and sample introduction. There are many varieties of covers to choose from, including one-piece, hinged, conical, and rotary style.