Cytiva's Whatman filter compressor can process six Mini-UniPrep syringeless filter devices simultaneously, resulting in a streamlined, cost-effective sample preparation process.
- Compatible with Mini UniPrep syringeless filter vials
- Eliminates the need for syringes, syringe filters, vials, and septa
- Compress six samples at once for minimal hand stress
- Polypropylene housing
The Mini-UniPrep 6 Position Compressor from Cytiva's business is used with Mini UniPrep syringeless filters
Preparing samples for use in an HPLC or UHPLC autosampler with syringe filters, vials, and septa requires substantial set-up time and space to manage the various consumables. Mini UniPrep syringeless filters cut multiple consumables out of the filtration process to save space and money, preparing samples in one-third the time required by other methods.
Add the 6-position multi-compressor to your process for even faster sample filtration with better ergonomics.
Browse the Mini UniPrep line for clear and amber versions, different membrane types, and slit or normal septa.