Carbon Dioxide Reagent Set, for CA-23 Test Kit, Hach
Proveedor: Hach
2437500
89231-072EA 1134.69 MXN
89231-072
Carbon Dioxide Reagent Set, for CA-23 Test Kit, Hach
Kits para test medioambiente
For determination of carbon dioxide by drop-count titration.
Información de suministro: Set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0.01 N).