SP Wilmad-LabGlass Hydroxypropoxy Determination Apparatus, SP Industries
Proveedor: SP Industries
LG-6606-100
14008-626EA 23121.07 MXN
14008-626
SP Wilmad-LabGlass Hydroxypropoxy Determination Apparatus, SP Industries
Componentes para destilación Sets de destilación
Complete apparatus as described in USP 23-NF 18 (421) for hydroxypropoxy group determinations.
  • A 250mL Erlenmeyer flask (C-4980-250) is supplied as a receiver
This borosilicate glass apparatus includes a 125mL conical bottom flask with a Standard Taper 24/40 outer joint, thermocouple well and an inlet with a 1.0mm capillary tip for nitrogen and water, distillation head, condenser head, and condenser.