Order Entry
BioClean™ Sterile Anti-Fog Spray, Ansell
  75972-514
undefined
BioClean™ Sterile Anti-Fog Spray, Ansell
  75972-514
 :  BAFS-601
$1,085.73
Select uom
Unit of Measure
Paquete de 10
  • Description:
    BioClean™ Sterile Anti-Fog Solution for Autoclavable Goggles
  • Size:
    60 ml
  • Cat. no.:
    75972-514
  • Supplier No.:
    BAFS-601