Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Proveedor: HiMedia Laboratories
M614-500G M614-5KG M614S-500G M614-2.5KG M614-500G M614-10KG M614-25KG M614-10KG M614S-500G M614-25KG M614-2.5KG
95059-662EA 24757.94 MXN
CA95059-662 95026-848 95059-662 CA95022-078 CA95022-076 95022-078 CA71002-740 95022-076 CA89130-344 71002-740 89130-344
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Medios Medios deshidratados para microbiología
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
  • Microbiological culture media
  • Packed in poly bottles