KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE®
292010-0000 292011-0000
KT292010-0000EA 4946.49 MXN
KT292010-0000 KT292011-0000
KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Aparatos de extracción Extractores Soxhlet y tipo Soxhlet
Soxhlet-type extraction apparatus is for use with extraction thimble, which has a coarse porosity.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.