VWR® Straight Sided Jars, Wide Mouth, HDPE
W311-1000
15900-292CS 87.02 MXN
15900-292
VWR® Straight Sided Jars, Wide Mouth, HDPE
Jars with foam-lined cap for use in metals analyses.
Certificaciones: Meet EPA contaminant-free sample containers specifications.
Información de suministro: Include certificate of analysis.
Envase: Custody-sealed.