Fluorescence Cells, PerkinElmer
Proveedor: Perkin Elmer
B0631113 B0631116 B0631107 B0631120 B0631113 B0631107 B0631110 B0631104 B0631104 B0631118 B0631116
97012-876PK 13643.37 MXN
45603-866 CA97012-876 97013-542 CA97012-880 45603-856 97012-876 CA97013-542 CA97013-548 97013-548 45603-870 97012-880
Fluorescence Cells, PerkinElmer
Cubetas Cubetas para espectrofotómetros
Standard rectangular macro and micro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
  • Clear windows for easy observation
  • Rectangular design
Quartz SUPRASIL® cells are available in a range of sizes for use with various sample volumes. All cells contain two clear windows each.