DPD Chlorine Reagent Powder Pop Dispenser, HF Scientific
Proveedor: HF Scientific
10504C 10506C 10446A 10448A 10445A
82003-184EA 2842.77 MXN
82003-186 82003-184 82003-189 97058-872 97058-874
DPD Chlorine Reagent Powder Pop Dispenser, HF Scientific
Kits para test medioambiente
This portable dispenser accurately delivers the correct volume of DPD reagent for free or total chlorine
  • Powder dissolves quickly in sample, leaving no messy residue behind
  • No spill dispensing into most round or square cuvettes
  • Dispenser design prevents clogging
To use, simply invert the dispenser over sample and press the button to dispense reagent.
Información de suministro: Supplied with enough DPD powder for 100 tests or in multi-packs providing 400 or 1000 tests.