VWR® Ice Buckets
Proveedor: VWR, part of Avantor
525130K 525130G 525070B 525130B
414004-246EA 2365.32 MXN
414004-249 414004-246 414004-245 414004-244
VWR® Ice Buckets
Durable rigid ice bucket with unbreakable lid for use with dry-ice and water-ice salt soution.
  • Excellent insulating properties
  • Resistant to temperatures from -20 to 70 °C