Open Handle Probes, Seedburo
Proveedor: Seedburo
22-OH 21-OH
56724-039EA 18191.16 MXN
56724-039 56724-038
Open Handle Probes, Seedburo
Muestreadores Muestreadores de sólidos
Open-ended probes without partitions are more convenient to empty and are widely used by the grain elevator operators for obtaining samples from truck loads.
  • Extra heavy gauge tubing for durable, time-tested sampling
  • Held upright and sample is dumped from the handle